Order Entry
United States
ContactUsLinkComponent
 
Ozone Instrumental Test 0 to 0.25 ppm
Catalog # 77674-846
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Ozone Instrumental Test 0 to 0.25 ppm
Catalog # 77674-846
Supplier:  CHEMetrics
Supplier Number:  K-7453
New Product
CAS Number:  

Specifications

  • Chemical method:
    Indigo
  • Application:
    Potable water;Bottled water;Food and beverages;Pharmaceutical solutions

Specifications

About this item

  • Analysis time: 1 minute Range: 0 to 0.25 ppm Method: Indigo
Delivery: The low range Ozone Vacu-vials® test kit (Indigo) comes in a cardboard box and contains everything needed to perform 30 tests: thirty Vacu-vials® ampoules, 25 ml sample cup, ampoule blank, and instructions. This test is designed for use in bottled water and drinking water applications where ozone is used as a disinfectant. This kit requires the use of a SAM photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell and with a Z dimension of 15 mm or less.