Paragon Scientific Distillation Temperature Check Standard - High Range
Catalog # 77975-565
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Distillation temperature check standard - high range
- Size:250 ml
- Storage Temperature:RT
Specifications
About this item
- Shipping conditions: Room temperature