TRC Nonidet P 40 Substitute
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:9016-45-9
Specifications
Frequently Bought Together
EZFlow® Syringe Filter, Sample Prep, PES, Foxx Life Sciences
Supplier: Foxx Life Sciences
Acrodisc® Syringe Filters for Prefiltration, Cytiva
Supplier: Cytiva
Diamond® PureFlow™ Nylon Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Syringe Filters, Non Sterile, Environmental Express®
Supplier: Environmental Express
Syringe Filters
Supplier: NEST SCIENTIFIC
SYRINGE FILTER PES STR 0.05UM 25MM PK100
Supplier: TISCH SCIENTIFIC MS
Target® Syringe Filters, PTFE, Thermo Scientific
Supplier: Thermo Scientific
Acrodisc® Syringe Filters, 25 mm
Supplier: Cytiva
MICROKROS 20CM 50K MPES 1MM MLL FLL STRL
Supplier: 0000019223
SYRINGE FILTER NYLON 0.22UM 13MM S CS75
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Supor® Membrane Disc Filters
Supplier: Cytiva
FILTER SYS 250ML PES 0.10UM 75MM S CS12
Supplier: VistaLab
Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
ACRODISC PSF WITH 0.2UM PVDF 50PK
Supplier: Cytiva
BOT TP FILT 500ML PES 0.45UM 90MM S CS24
Supplier: VistaLab
Whatman™ Polydisc AS In-Line Filters, Whatman products (Cytiva)
Supplier: Cytiva