Order Entry
United States
ContactUsLinkComponent
Dr. Ehrenstorfer EPA Method 537 Internal Standard Primary Dilution Standard 1-4 µg/ml in Methanol:Water
Dr. Ehrenstorfer EPA Method 537 Internal Standard Primary Dilution Standard 1-4 µg/ml in Methanol:Water
Catalog # 77976-058
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Dr. Ehrenstorfer EPA Method 537 Internal Standard Primary Dilution Standard 1-4 µg/ml in Methanol:Water
Catalog # 77976-058
Supplier:  LGC Standards
Supplier Number:  DRE-A30000093MW
New Product
CAS Number:  

Specifications

  • Matrix:
    Methanol:Water 96:4
  • Description:
    EPA Method 537 Internal standard primary dilution standard 1 - 4 µg/ml in Methanol:Water
  • Size:
    1.2 ml
  • Shipping Conditions:
    20 °C
  • Storage Temperature:
    −18 °C
  • Certification:
    ISO 17034

Specifications

About this item