Paragon Scientific Density 50 °C (Nominal: 0.8659 g/ml @ 50 °C)
New Product
- Shipping conditions: Room temperature
Frequently Bought Together
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
Captiva Syringe Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Outer Tube-Sets for Semi and Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Direct Connect Liners for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tools for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
Pump Valves and Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Split Liners for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)