ContactUsLinkComponent
Chloride Titration Test 250 to 2500 ppm
Chloride Titration Test 250 to 2500 ppm
  77674-746
New Product
 :  
undefined
Chloride Titration Test 250 to 2500 ppm
  77674-746
 :  K-2051
New Product
 :  
  • Chemical method:
    Ferric thiocyanate
  • Application:
    Drinking water;Surface and saline water;Domestic and industrial waste water
Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE
  • Range: 250 to 2500 ppm MDL: 250 ppm Method: Mercuric nitrate Increments: 250, 275, 300, 325, 375, 400, 450, 500, 625, 750, 875, 1000, 1250, 1750, 2500 ppm
 : Kit K-2051 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, activator solution, titrettor, 25 ml sample cup, and instructions.