Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Density 25 °C (Nominal: 0.7730 g/ml @ 25 °C)
Paragon Scientific Density 25 °C (Nominal: 0.7730 g/ml @ 25 °C)
Catalog # 77975-685
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Density 25 °C (Nominal: 0.7730 g/ml @ 25 °C)
Catalog # 77975-685
Supplier:  LGC Standards
Supplier Number:  ALK-DEN25-03-500ML
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Density 25 °C (Nominal: 0.7730 g/ml @ 25 °C), Standard type: Multi-component
  • Size:
    500 ml

Specifications

About this item

  • Shipping conditions: Room temperature