Order Entry
United States
Orders LinkContactUsLinkComponent
TRC Hexanal

TRC Hexanal

Hexanal, 50 g
Catalog # 77985-910
New Product
Supplier:   LGC Standards
Supplier #:  TRC-H292130-50G
TRC Hexanal
Catalog # 77985-910
Supplier:  LGC Standards
New Product
CAS Number:  
TRC Hexanal
TRC Hexanal

Hexanal, 50 g

CAS Number:  
Price
$177.00
Log in for availability
Each (0.02g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications

Specifications

  • Description:
    Hexanal
  • Size:
    50 g
  • Cat. No.:
    77985-910
  • CAS No.:
    66-25-1

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.