Order Entry
United States
ContactUsLinkComponent
 
Paragon Scientific Refractive Index (Nominal: 1.4023 @ 25 °C), Certified @ 20, 25 and 30 °C
Catalog # 77975-905
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Refractive Index (Nominal: 1.4023 @ 25 °C), Certified @ 20, 25 and 30 °C
Catalog # 77975-905
Supplier:  LGC Standards
Supplier Number:  ALK-PS-RI-03K
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Refractive index (nominal: 1.4023 @ 25 °C), certified @ 20, 25 and 30 °C
  • Size:
    5×10 ml
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component

    Shipping conditions: Room temperature