TRC Dasatinib
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:302962-49-8
Specifications
Frequently Bought Together
Whatman™ Puradisc Syringe Filters, CA, Whatman products (Cytiva)
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
Whatman™ ReZist Air Venting Filter, Whatman products (Cytiva)
Supplier: Cytiva
Whatman™ GD/XP Syringe Filters
Supplier: Cytiva
Whatman™ Polydisc AS In-Line Filters, Whatman products (Cytiva)
Supplier: Cytiva
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva
Titan3™ Nylon (Polyamide) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
FILTER SYRINGE PTFE 0.2UM POR 25MM PK100
Supplier: Cole Parmer
LabExact® PTFE Inline Filters, Sterile¹/₄ to ³/₈" Barb Ports
Supplier: I.W. TREMONT CO., INC.
SYRINGE FILTER PES STR 0.05UM 25MM PK100
Supplier: TISCH SCIENTIFIC MS
MICRO 20CM 0.65UM MPES 0.75MM
Supplier: 0000019223
BOT TP FILT 150ML PES 0.45UM 50MM S CS24
Supplier: VistaLab
Diamond® PureFlow™ PTFE Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
FILTER SYS 150ML PES 0.45UM 50MM S CS12
Supplier: VistaLab
Vacuum Pumps, Sartorius
Supplier: Sartorius
Syringe Filters, Argos Technologies
Supplier: Antyila Scientific