Paragon Scientific European Pharmacopoeia Reagent Coloration - Primary Solution Red
Catalog # 77975-502
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:European pharmacopoeia reagent coloration - Primary solution red
- Size:100 ml
- Storage temperature:RT