Paragon Scientific Density 25 °C (Nominal: 0.6564 g/ml @ 25 °C)
New Product
- Shipping conditions: Room temperature
Frequently Bought Together
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for VacElut Cartridge Manifolds, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
Accessories for Stay Safe Caps for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Cylinder Supplies, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tools for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Autosampler Supplies, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)