Paragon Scientific Small Sample Viscosity (3420 cSt @ 40 °C), Certified @ 40 and 100 °C
Catalog # 77975-778
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Small sample viscosity (3420 cSt @ 40 °C), certified @ 40 and 100 °C
- Size:60 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component