Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Relative Density 15 °C (Nominal: 0.7183 @ 15 °C)
Paragon Scientific Relative Density 15 °C (Nominal: 0.7183 @ 15 °C)
Catalog # 77975-944
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Relative Density 15 °C (Nominal: 0.7183 @ 15 °C)
Catalog # 77975-944
Supplier:  LGC Standards
Supplier Number:  ALK-RDEN15-02
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Relative density 15 °C (Nominal: 0.7183 @ 15 °C), Standard type: Single component
  • Size:
    500 ml

Specifications

About this item

  • Shipping conditions: Room temperature