Paragon Scientific Type PSU1600, Viscosity Reference Standard
77975-924
New Product
:
- Description:Type PSU1600, viscosity reference standard
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component