Paragon Scientific Type CL200 (CL20), Cold Crank Simulator Viscosity
77975-532
New Product
:
- Description:Type CL200 (CL20), cold crank simulator viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component