$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
TRC Forchlorfenuron
TRC Forchlorfenuron
77985-059
:
LGC Standards
New Product
:
TRC Forchlorfenuron
77985-059
:
LGC Standards
:
TRC-F691050-100MG
New Product
:
CAS No.:
68157-60-8
Description:
Forchlorfenuron
Size:
100 mg