ContactUsLinkComponent
Hardness Titration Test Kit 50 to 500 ppm
  77674-738
New Product
 :  
undefined
Hardness Titration Test Kit 50 to 500 ppm
  77674-738
 :  K-1705
New Product
 :  
  • Chemical method:
    EDTA (total)
  • Application:
    Drinking water;Surface water;Saline water
This kit uses the EGTA chemistry to measure the hardness of water by measuring the dissolved calcium in the water.
  • Range: 50 to 500 ppm as CaCO₃ MDL: 50 ppm Method: EGTA (calcium) Increments: 50, 55, 60, 65, 70, 75, 80, 90, 100, 125, 150, 175, 200, 250, 350, 500 ppm CaCO₃
Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure — no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE
 : The K-1705 hardness calcium Titrets® test kit (50 to 500 ppm) comes in a cardboard box and contains everything needed to perform 30 tests: thirty Titrets® ampoules with valve assemblies, indicator solution, plastic titrettor, 25 ml sample cup, and instructions.