Dr. Ehrenstorfer UK Drinking Water Inspectorate (DWI) PFAS-48 Mixture B 10 µg/ml in Methanol:Water
Catalog # 77976-054
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- Description:UK Drinking water inspectorate (DWI) PFAS - 48 mixture B 10 µg/ml in Methanol:Water
- Size:1 ml
- Shipping Conditions:20 °C
- Storage Temperature:−18 °C
- Certification:ISO 17034