TRC Methotrexate-d3 (Technical Grade) (100 µg/ml in Methanol with 1% 0.1N NaOH)
77987-715
New Product
:
- Matrix:Methanol:NaOH
- CAS No.:432545-63-6
- Description:Methotrexate-d3 (Technical Grade) (100 µg/ml in Methanol with 1% 0.1N NaOH)
- Size:5×1 ml