Dr. Ehrenstorfer Tris(1H,1H,2H,2H-perfluorooctyl)phosphate
77976-069
New Product
:
- CAS No.:165325-62-2
- Description:Tris(1H, 1H, 2H, 2H - perfluorooctyl)phosphate
- Size:10 mg
- Shipping Conditions:20 °C
- Formula:C₂₄H₁₂F₃₉O₄P
- Formula Weight:1136,261 g/mol
- Storage Temperature:4 °C
- Certification:ISO/IEC 17025