Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Color Standard USP (631) Cupric Sulfate CS
Paragon Scientific Color Standard USP (631) Cupric Sulfate CS
Catalog # 77976-030
Supplier:  LGC Standards
New Product
CAS Number:  
Paragon Scientific Color Standard USP (631) Cupric Sulfate CS
Catalog # 77976-030
Supplier:  LGC Standards
Supplier Number:  ALK-USPPRBL
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Color standard USP (631) Cupric Sulfate CS
  • Size:
    100 ml
  • Storage temperature:
    RT

Specifications

About this item

  • Matrix: Aq. HCl