$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
TRC N-(Demethyl Formate) Oxibendazole Hydrochloride
TRC N-(Demethyl Formate) Oxibendazole Hydrochloride
Catalog # 77981-457
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC N-(Demethyl Formate) Oxibendazole Hydrochloride
Catalog # 77981-457
Supplier:
LGC Standards
Supplier Number:
TRC-D231645-100MG
New Product
CAS Number:
Specifications
CAS No.:
1538624-34-8
Description:
N-(Demethyl Formate) Oxibendazole Hydrochloride
Size:
100 mg
Specifications
About this item
Product Details
Product Details