Dr. Ehrenstorfer EPA Method 533 Isotope Performance Standard 1-3 µg/ml in Methanol:Water
Catalog # 77976-057
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- Description:EPA Method 533 Isotope performance standard 1 - 3 µg/ml in Methanol:Water
- Size:1.2 ml
- Shipping Conditions:20 °C
- Storage Temperature:−18 °C
- Certification:ISO 17034