Paragon Scientific Density 20 °C (Nominal: 0.6609 g/ml @ 20 °C)
New Product
- Shipping conditions: Room temperature
Frequently Bought Together
CrossLab Instrument Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
ADM Flow Meter, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Maintenance Kits for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing and Fittings for 8453/8454 UV Systems
: AGILENT TECHNOLOGIES, INC (CSD)
Flow Through Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
pH Gradient Buffers
: Thermo Scientific
Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Intuvo Flow Chips, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Split Liners for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)