Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Silicone Fluid 5 cSt @ 25 °C (for use 20 to 60 °C), Viscosity Bath Media
Paragon Scientific Silicone Fluid 5 cSt @ 25 °C (for use 20 to 60 °C), Viscosity Bath Media
Supplier:  LGC Standards
New Product
Certificates
undefined
Paragon Scientific Silicone Fluid 5 cSt @ 25 °C (for use 20 to 60 °C), Viscosity Bath Media
Supplier:  LGC Standards
New Product

Specifications

  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT

Specifications

Frequently Bought Together