TRC Ceftiofur Hydrochloride
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:103980-44-5
Specifications
Frequently Bought Together
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
SYRINGE FILTER PTFE N-ST 5UM 25MM PK100
Supplier: TISCH SCIENTIFIC MS
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva
Diamond® PureFlow™ PTFE Syringe Filters, Non Sterile, Globe Scientific
Supplier: Globe Scientific
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Supplier: Cytiva
Minisart® Acticosart Syringe Filters, Sartorius
Supplier: Sartorius
CellPro™ Syringe Filters
Supplier: Alkali Scientific
Acrodisc® Syringe Filters for Prefiltration, Cytiva
Supplier: Cytiva
Diamond® PureFlow™ PTFE Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
Whatman™ Polydisc AS In-Line Filters, Whatman products (Cytiva)
Supplier: Cytiva
SunFlo™ Dissolution Filters, SUN-SRi
Supplier: Thermo Scientific
BOT TP FILT PP 1000ML NYLN .45UM NS CS12
Supplier: VistaLab