Dr. Ehrenstorfer Korea cosmetics testing PFAS Mixture 1 µg/ml in Methanol:Water
Catalog # 77976-051
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- Description:Korea cosmetics testing PFAS mixture 1 µg/ml in Methanol:Water
- Size:1 ml
- Shipping Conditions:20 °C
- Storage Temperature:−18 °C
- Certification:ISO/IEC 17025