Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Viscosity Set, Cold Crank Simulator: Type CL14, CL19, CL22, CL25, CL28, CL32, CL48, CL68
Paragon Scientific Viscosity Set, Cold Crank Simulator: Type CL14, CL19, CL22, CL25, CL28, CL32, CL48, CL68
Catalog # 77976-035
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Viscosity Set, Cold Crank Simulator: Type CL14, CL19, CL22, CL25, CL28, CL32, CL48, CL68
Catalog # 77976-035
Supplier:  LGC Standards
Supplier Number:  ALK-VIS-CCS-8
New Product
CAS Number:  

Specifications

  • Description:
    Viscosity set, cold crank simulator type: CL14, CL19, CL22, CL25, CL28, CL32, CL48, CL68
  • Size:
    8×500 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component