TRC Benzyldimethyltetradecylammonium Chloride
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:139-08-2
Specifications
Frequently Bought Together
EZFlow® Syringe Filter, CA, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Luer Lock Stop Cocks
Supplier: S17 SCIENCE LLC.
J.T.Baker®, Syringe Filters
Supplier: Avantor
Whatman™ GD/XP Syringe Filters
Supplier: Cytiva
FILTER SYS 1000ML PES 0.10UM 90MM S CS12
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Minisart® Syringe Filters NML
Supplier: Sartorius
MICRO 20CM 0.65UM MPES 0.75MM
Supplier: 0000019223
BOT TP FILT 150ML PES 0.45UM 50MM S CS24
Supplier: VistaLab
FILTER SYS 150ML PES 0.10UM 50MM S CS12
Supplier: VistaLab
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva