Paragon Scientific Relative Density 25 °C (Nominal: 0.7101 @ 25 °C)
Catalog # 77975-957
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Relative density 25 °C (Nominal: 0.7101 @ 25 °C), Standard type: Single component
- Size:500 ml
Specifications
About this item
- Shipping conditions: Room temperature