Paragon Scientific Pt-Co/Hazen/APHA (Nominal: 50), Color
Catalog # 77975-940
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Pt-Co/Hazen/APHA (nominal: 50), color
- Size:500 ml
- Storage temperature:RT