Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Ethanol, 70% ABV (Alcohol by Volume)
Paragon Scientific Ethanol, 70% ABV (Alcohol by Volume)
Catalog # 77975-730
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Ethanol, 70% ABV (Alcohol by Volume)
Catalog # 77975-730
Supplier:  LGC Standards
Supplier Number:  ALK-ETWA70
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Ethanol, 70% ABV (Alcohol by Volume)
  • Size:
    25 ml

Specifications

About this item

  • Shipping condition: Room temperature