Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Refractive Index (Nominal: 1.4206 @ 25 °C), Certified @ 20, 25 and 30 °C
Paragon Scientific Refractive Index (Nominal: 1.4206 @ 25 °C), Certified @ 20, 25 and 30 °C
  77975-908
New Product
 :  
undefined
Paragon Scientific Refractive Index (Nominal: 1.4206 @ 25 °C), Certified @ 20, 25 and 30 °C
  77975-908
 :  ALK-PS-RI-05K
New Product
 :  

 

  • Pack type:
    Glass bottle
  • Description:
    Refractive index (nominal: 1.4206 @ 25 °C), certified @ 20, 25 and 30 °C
  • Size:
    5×10 ml
  • Certification:
    ISO 17034

 

 

  • Standard type: Single component

    Shipping conditions: Room temperature