Order Entry
United States
ContactUsLinkComponent
Oxygen Instrumental Test 0 to 1 ppm
Oxygen Instrumental Test 0 to 1 ppm
Catalog # 77674-848
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Oxygen Instrumental Test 0 to 1 ppm
Catalog # 77674-848
Supplier:  CHEMetrics
Supplier Number:  K-7553
New Product
CAS Number:  

Specifications

  • Chemical method:
    Rhodazine D
  • Application:
    Boiler water;Cooling water;Sea water;Power generation;Petroleum refining;Water treatment

Specifications

About this item

  • Range: 0 to 1.000 ppm Method: Rhodazine D
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, sampling tube, permanent mounting clamp, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.