Paragon Scientific Relative Density 15 °C (Nominal: 0.8111 @ 15 °C)
77975-946
New Product
:
- Pack type:Glass bottle
- Description:Relative density 15 °C (Nominal: 0.8111 @ 15 °C), Standard type: Single component
- Size:500 ml
- Shipping conditions: Room temperature