$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
TRC 10-trans,12-cis-Linoleic Acid
TRC 10-trans,12-cis-Linoleic Acid
Catalog # 77988-044
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 10-trans,12-cis-Linoleic Acid
Catalog # 77988-044
Supplier:
LGC Standards
Supplier Number:
TRC-L462520-50MG
New Product
CAS Number:
Specifications
CAS No.:
2420-56-6
Description:
10-trans,12-cis-Linoleic Acid
Size:
50 mg
Specifications
About this item
Product Details
Product Details