Paragon Scientific Type S60S, High Temperature Viscosity
Catalog # 77976-000
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type S60S, High temperature viscosity emperature viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component