Lipomed 25 mg/dL Aq. Ethanol Standard Solution, 25 mg/dL in Water
Catalog # 77976-156
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Water
- CAS No.:64-17-5
- Description:25 mg/dL Aq. Ethanol Standard Solution, 25 mg/dL in Water
- Size:5×1 ml