Paragon Scientific Type CL300 (CL30), Cold Crank Simulator Viscosity
77975-536
New Product
:
- Description:Type CL300 (CL30), cold crank simulator viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component