ContactUsLinkComponent
Fungus GMS Control Slides
Fungus GMS Control Slides
Catalog # 77907-906
Supplier:  StatLab
New Product
undefined
Fungus GMS Control Slides
Catalog # 77907-906
Supplier:  StatLab
Supplier Number:  CS-FUNGUS/GMS/25
New Product
Specifications
  • Description:
    Fungus GMS control slides
Specifications
About this item
Fungus GMS control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes 1 stained slide
    Tissue: known positive
    Special stain or antibody: GMS stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.