ContactUsLinkComponent
Mucin (Human) Control Slides
Mucin (Human) Control Slides
Catalog # 77907-944
Supplier:  StatLab
New Product
undefined
Mucin (Human) Control Slides
Catalog # 77907-944
Supplier:  StatLab
Supplier Number:  CS-MUCI/25
New Product
Specifications
  • Description:
    Mucin (Human) control slides
Specifications
About this item
Mucin (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
    Tissue: known positive
    Special stain or antibody: Mucicarmine stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.