ContactUsLinkComponent
Herpes Type 1 IHC Control Slide
Herpes Type 1 IHC Control Slide
Catalog # 77907-916
Supplier:  StatLab
New Product
undefined
Herpes Type 1 IHC Control Slide
Catalog # 77907-916
Supplier:  StatLab
Supplier Number:  CSH0425P
New Product
Specifications
  • Description:
    Herpes Type 1 IHC control slides
Specifications
About this item
Herpes Type 1 IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
  • Tissue: known positive
  • Special stain or antibody: Herpes simplex virus I
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.