Mesothelioma IHC Control Slide
77907-934
: StatLab
New Product
- Description:Mesothelioma IHC control slides
Mesothelioma IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Stained slide is not included
- Tissue: Mesothelioma
- Special stain or antibody: Mesothelioma
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.