Nocardia Control Slide
77907-946
: StatLab
New Product
- Description:Nocardia control slides
Nocardia control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Stained slide is not included
- Tissue: known positive
- Special stain or antibody: Fite's stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.