ContactUsLinkComponent
Herpes Simplex Virus, Type 1 and 2 IHC Control Slides
Herpes Simplex Virus, Type 1 and 2 IHC Control Slides
Catalog # 77907-926
Supplier:  StatLab
New Product
undefined
Herpes Simplex Virus, Type 1 and 2 IHC Control Slides
Catalog # 77907-926
Supplier:  StatLab
Supplier Number:  CS-HSV1/2/25
New Product
Specifications
  • Description:
    Herpes Simplex Virus, Type 1 and 2 IHC control slides
Specifications
About this item
One slide in each box features a serial section that has been stained using the specified staining procedure that is included in each box.