ContactUsLinkComponent
Cincinnati Surgical Blade Handle, #4
Cincinnati Surgical Blade Handle, #4
Catalog # 77907-922
Supplier:  StatLab
New Product
Cincinnati Surgical Blade Handle, #4
Catalog # 77907-922
Supplier:  StatLab
Supplier Number:  CS-H4
New Product
Specifications
  • Description:
    Cincinnati surgical blade handle
  • Blade no.:
    #4
Specifications
About this item
Manufactured from special cold-formed nickel steel.
  • Handles are stainless steel
  • Non sterile
Each handle is designed to incorporate a satin finish, smooth loading, and balance. Cincinnati surgical blade handles are designed for use with Cincinnati surgical blades. It's important to ensure that the right blade is paired with the right handle.