ContactUsLinkComponent
Spirochete Control Slides
Spirochete Control Slides
Catalog # 77907-974
Supplier:  StatLab
New Product
undefined
Spirochete Control Slides
Catalog # 77907-974
Supplier:  StatLab
Supplier Number:  CS-SPIRO/25
New Product
Specifications
  • Description:
    Spirochete control slides
Specifications
About this item
Spirochete control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
    Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.