HistoBond®+ Supa Mega White Charged Microscope Slide
77908-080
: StatLab
New Product
- Description:HistoBond®+ supa mega white charged microscope slides
- L×W:52×76 mm
The autoclavable HistoBond®+ Supa Mega White Charged Slides are supplied pre-cleaned and ready for use in cardboard boxes of 50 slides.
- Suitable for problem tissue
- Suitable for IHC
- White printed marking area
- Pre-cleaned and ready to use
- Time-saving option
- Manufactured to ISO 8037/1
The HistoBond®+ Supa Mega White Charged Microscope Slide is made of soda-lime glass and ensures excellent specimen adhesion. The provision of a white marking area means slides can be identified with a pen, pencil, or label.