Ambrisentan Impurity 10, TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:177036-78-1
Specifications
Frequently Bought Together
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Supplier: Thermo Scientific Chemicals
Water, extra pure, deionized
Supplier: Thermo Scientific Chemicals
R-BAIBA 95%
Supplier: Ambeed
Trichloroethylene 99.6% stabilized ACS
Supplier: Thermo Scientific Chemicals
Hydrofluoric acid 48 - 51% in water for analysis
Supplier: Thermo Scientific Chemicals
(R)-Ibuprofen 96%
Supplier: Ambeed
Chloroform 99.9% stabilized, ECD tested for residue analysis, for pesticide residue analysis residue <0.0005%
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Pyridine 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
Acetone 99+%, extra pure
Supplier: Thermo Scientific Chemicals
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
p-Xylene 99% for HPLC
Supplier: Thermo Scientific Chemicals
Tetrahydrofuran 99+% stabilized, extra pure
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane 99.8+%, Extra Pure
Supplier: Thermo Scientific Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Scientific Chemicals
N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals